Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Idgf5
Entrez Gene ID
37104
Gene Name
Imaginal disc growth factor 5
Gene Aliases
CG5154, Cht14, DmIDGF5, Dmel\CG5154, Dmel_CG5154, IDGF, IDGF-5, IDGF5, idgf5
Chromosome Location
Chr.2R: 18393666 - 18395282 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0064237
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137477.4 NP_611321.3 2 - 3 734 98
GenBank mRNA BT012501.1 - 2 - 3 711 98
BT044434.1 - 2 - 3 722 98

View Full Product Details