Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jheh1
Entrez Gene ID
251984
Gene Name
Juvenile hormone epoxide hydrolase 1
Gene Aliases
142196_at, BEST:GH06241, CG15101, Dmel\CG15101, Dmel_CG15101, JH, JHEH, Jheh
Chromosome Location
Chr.2R: 18853434 - 18855087 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010053
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_137541.4 NP_611385.1 1 - 2 241 70
GenBank mRNA AF517545.1 - 1 - 2 207 70
BT001753.1 - 1 - 2 264 70

View Full Product Details