Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arl6
Entrez Gene ID
37236
Gene Name
ADP ribosylation factor-like 6
Gene Aliases
A1ZBK9, ARL6, BBS3, CG7735, Dmel\CG7735, Dmel_CG7735
Chromosome Location
Chr.2R: 19444179 - 19445058 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0034446
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299686.1 NP_001286615.1 2 - 3 350 76
NM_137577.3 NP_611421.1 2 - 3 353 76

View Full Product Details