Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CoRest
Entrez Gene ID
32941
Gene Name
CG42687-PA
Gene Aliases
BcDNA:GH01486, BcDNA:LD11215, CG32534, CG32535, CG33525, CG3878, CG42687, CoREST, CoRESTG, Dmel\CG42687, Dmel_CG42687, dCoREST, i169
Chromosome Location
Chr.X: 19521929 - 19530896 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261573
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014752.4 NP_001014752.2 3 - 4 655 56
NM_001014753.3 NP_001014753.1 3 - 4 655 56
NM_001014754.1 NP_001014754.1 4 - 5 616 56
NM_001014755.2 NP_001014755.1 3 - 4 901 56
NM_001014756.2 NP_001014756.1 3 - 4 655 56
NM_001201742.3 NP_001188671.1 3 - 4 655 56
NM_001258824.1 NP_001245753.1 3 - 4 735 56
NM_001258825.2 NP_001245754.1 3 - 4 835 56
NM_001272777.1 NP_001259706.1 3 - 4 663 56
GenBank mRNA BT023817.1 - 3 - 4 633 56
BT099711.1 - 3 - 4 559 56

View Full Product Details