Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ns3
Entrez Gene ID
31097
Gene Name
Nucleostemin 3
Gene Aliases
A60, BACN32G11.5, CG14788, Dmel\CG14788, Dmel_CG14788, EG:BACN32G11.5, dNS3, dmNuc3, l(1)2Ad, l(1)BA11, l(1)G0431, ns3
Chromosome Location
Chr.X: 1461844 - 1464220 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266284
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297850.1 NP_001284779.1 1 - 2 209 67
NM_130559.3 NP_569915.1 1 - 2 209 67
GenBank mRNA AY047556.1 - 1 - 2 178 67

View Full Product Details