Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Oatp58Db
- Entrez Gene ID
-
37544
- Gene Name
- Organic anion transporting polypeptide 58Db
- Gene Aliases
-
58Db, CG3382, Dmel\CG3382, Dmel_CG3382, oatp 58Db
- Chromosome Location
-
Chr.2R: 22221180 - 22223499 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034715
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_137814.4 |
NP_611658.3 |
1 - 2 |
176 |
86 |
View Full Product Details