Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NtR
Entrez Gene ID
43935
Gene Name
CG6698-PA
Gene Aliases
CG6698, DNtR, Dmel\CG6698, Dmel_CG6698, anon-WO0138359.3
Chromosome Location
Chr.2R: 22127808 - 22130184 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029147
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259528.2 NP_001246457.1 5 - 6 1356 75
NM_143701.4 NP_651958.2 5 - 6 1377 75

View Full Product Details