Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ND-PDSW
Entrez Gene ID
44228
Gene Name
NADH dehydrogenase (ubiquinone) PDSW subunit
Gene Aliases
BcDNA:RE65754, CG8844, Dmel\CG8844, Dmel_CG8844, NIDM, PDSW, Pdsw, anon-EST:Posey91, l(2)k10101, pdsw
Chromosome Location
Chr.2L: 3466157 - 3467334 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0021967
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_143715.3 NP_651972.1 2 - 3 170 90
NM_164537.2 NP_722908.1 1 - 2 257 90
GenBank mRNA AY060865.1 - 2 - 3 132 90
AY113547.1 - 2 - 3 166 90

View Full Product Details