Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vps28
Entrez Gene ID
47408
Gene Name
Vacuolar protein sorting 28
Gene Aliases
CG12770, DmVps28, Dmel\CG12770, Dmel_CG12770, Dvps28, ESCRT-I, ESCRT-II, dVps28, dvps28, l(2)k16503, vps28
Chromosome Location
Chr.2R: 8078869 - 8080041 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0021814
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273867.2 NP_001260796.1 1 - 1 750 103

View Full Product Details