Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
drpr
Entrez Gene ID
38218
Gene Name
draper
Gene Aliases
BcDNA:GH03529, CG18172, CG2086, CT41022, CT6730, DRPR, Dmel\CG2086, Dmel_CG2086, Draper, Draper I, Drpr
Chromosome Location
Chr.3L: 1715595 - 1731107 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0027594
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274346.1 NP_001261275.1 8 - 9 2266 73
NM_001274347.1 NP_001261276.1 10 - 11 3553 73
NM_058102.3 NP_477450.1 8 - 9 2209 73
GenBank mRNA BT044170.1 - 8 - 9 2195 73

View Full Product Details