Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr64b
- Entrez Gene ID
-
117480
- Gene Name
- Gustatory receptor 64b
- Gene Aliases
-
CG14986, CG32257, Dmel64b, Dmel\CG32257, Dmel_CG32257, GR64b, Gr64, anon-WO0118547.604
- Chromosome Location
-
Chr.3L: 4028605 - 4032032 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045478
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168049.2 |
NP_728921.1 |
5 - 6 |
1116 |
61 |
- Gene Symbol
- Gr64c
- Entrez Gene ID
-
117479
- Gene Name
- Gustatory receptor 64c
- Gene Aliases
-
CG14986, CG32256, Dmel64c, Dmel\CG32256, Dmel_CG32256, GR64c, Gr64, gr64c
- Chromosome Location
-
Chr.3L: 4028605 - 4032032 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0045477
View Full Product Details