Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ero1L
- Entrez Gene ID
-
38500
- Gene Name
- Endoplasmic reticulum oxidoreductin-1-like
- Gene Aliases
-
CG1333, Dmel\CG1333, Dmel_CG1333, ERO1-L, Ero1, Ero1l, ero1L, kiga, l(3)64Ag, l(3)SH22
- Chromosome Location
-
Chr.3L: 4170404 - 4173789 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261274
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139608.4 |
NP_647865.2 |
2 - 3 |
656 |
96 |
GenBank mRNA |
BT021422.1 |
- |
2 - 3 |
604 |
96 |
View Full Product Details