Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sif
Entrez Gene ID
43892
Gene Name
still life
Gene Aliases
CG32414, CG34418, CG5256, CG5406, Dm_3L:26493, Dmel\CG34418, Dmel_CG34418, RhoGEF, SIF, Sif, Stilllife, lincRNA.447, slf
Chromosome Location
Chr.3L: 5657913 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085447
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104047.5 NP_001097517.1 12 - 13 3590 64
NM_001104048.2 NP_001097518.2 14 - 15 3250 64
NM_001104049.4 NP_001097519.2 13 - 14 3045 64
NM_001274517.3 NP_001261446.1 13 - 14 3647 64
NM_001274519.3 NP_001261448.1 13 - 14 3620 64
NM_001300018.2 NP_001286947.1 12 - 13 4602 64
NM_001347763.1 NP_001334674.1 14 - 15 3566 64
NM_001347764.1 NP_001334675.1 14 - 15 3602 64
NM_168126.4 NP_729084.1 12 - 13 2600 64

Gene Symbol
CG46320
Entrez Gene ID
30522409
Gene Name
CG46320-PA
Gene Aliases
Dmel\CG46320, Dmel_CG46320, sif-5'utr
Chromosome Location
Chr.3L: 5664121 - 5749599 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0284236
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001347801.1 NP_001334709.1 12 - 13 3590 64
NM_001347802.1 NP_001334710.1 14 - 15 3566 64

View Full Product Details