Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or65c
- Entrez Gene ID
-
318013
- Gene Name
- Odorant receptor 65c
- Gene Aliases
-
65c, CG32403, DOR65, Dmel\CG32403, Dmel_CG32403, LU27.3
- Chromosome Location
-
Chr.3L: 6327608 - 6329069 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041623
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168165.2 |
NP_729163.2 |
1 - 2 |
113 |
81 |
View Full Product Details