Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- hdm
- Entrez Gene ID
-
31705
- Gene Name
- hold'em
- Gene Aliases
-
CG15329, Dmel\CG15329, Dmel_CG15329
- Chromosome Location
-
Chr.X: 7918517 - 7920131 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0029977
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132194.1 |
NP_572422.1 |
1 - 2 |
918 |
88 |
View Full Product Details