Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dpr20
- Entrez Gene ID
-
38101
- Gene Name
- defective proboscis extension response 20
- Gene Aliases
-
CG12191, CT9818, Dmel\CG12191, Dmel_CG12191, Dpr-20, Dpr20
- Chromosome Location
-
Chr.3L: 852720 - 857248 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035170
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_138222.3 |
NP_612066.1 |
3 - 4 |
1476 |
83 |
GenBank mRNA |
AY051512.1 |
- |
3 - 4 |
1455 |
83 |
View Full Product Details