Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oseg4
Entrez Gene ID
38219
Gene Name
Outer segment 4
Gene Aliases
CG2069, Dmel\CG2069, Dmel_CG2069, IFT121, OSEG, OSEG4, WDR35, Wdr35, cytC40, oseg4
Chromosome Location
Chr.3L: 1730993 - 1735767 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035264
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274349.1 NP_001261278.1 5 - 6 1499 88
NM_139396.4 NP_647653.1 5 - 6 1499 88
GenBank mRNA AY061373.1 - 5 - 6 1358 88

View Full Product Details