Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sphinx1
- Entrez Gene ID
-
318005
- Gene Name
- CG32383-PB
- Gene Aliases
-
CG32383, CG8555, Dmel\CG32383, Dmel_CG32383, SPH194, SPH194a, Sphinx-1, Sphinx1, Spx1, sphinx-1
- Chromosome Location
-
Chr.3L: 7438421 - 7439405 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052383
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168214.2 |
NP_729255.2 |
1 - 1 |
67 |
105 |
View Full Product Details