Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sarm
- Entrez Gene ID
-
38895
- Gene Name
- Sterile alpha and Armadillo motif
- Gene Aliases
-
CG13680, CG13681, CG34373, CG43119, CG7905, CG7915, Dmel\CG43119, Dmel_CG43119, Ect4, Ect4/SARM, SARM, anon-WO0118547.272, dSARM, dSARM1, dSarm, dsarm, ect4
- Chromosome Location
-
Chr.3L: 8063874 - 8108897 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0262579
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168254.2 |
NP_729327.1 |
2 - 3 |
925 |
64 |
GenBank mRNA |
AY075346.1 |
- |
2 - 3 |
906 |
64 |
View Full Product Details