Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Ptip
- Entrez Gene ID
-
39552
- Gene Name
- PAX transcription activation domain interacting protein
- Gene Aliases
-
CG32133, CG6532, CG8797, Dmel\CG32133, Dmel_CG32133, PTIP, ptip
- Chromosome Location
-
Chr.3L: 14020315 - 14029121 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0052133
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168571.3 |
NP_729947.2 |
6 - 7 |
6559 |
61 |
View Full Product Details