Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pka-C3
- Entrez Gene ID
-
39733
- Gene Name
- Protein kinase, cAMP-dependent, catalytic subunit 3
- Gene Aliases
-
5-23, CG6117, DC2, Dmel\CG6117, Dmel_CG6117, Dpck, PKA, PKA C3, PKA-C-3, PKA-C3, PKAC3, Pk72A, Pk?5, pka-C3
- Chromosome Location
-
Chr.3L: 15925805 - 15954797 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000489
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_168638.3 |
NP_730083.2 |
2 - 3 |
211 |
66 |
View Full Product Details