Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mbs
Entrez Gene ID
49070
Gene Name
Myosin binding subunit
Gene Aliases
0573/06, 0953/04, CG32156, CG5600, CG5891, DMBS, DMYPT, DMbs, Dmel\CG32156, Dmel_CG32156, MBS, Mypt, l(3)03802, l(3)72Dd, l(3)S005331b, l(3)S057306, l(3)S057306b, l(3)S095304, l(3)j3B4, l(3)j7A1, mbs
Chromosome Location
Chr.3L: 16051959 - 16082026 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005536
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104144.3 NP_001097614.1 10 - 11 2152 62
NM_001104145.3 NP_001097615.1 11 - 12 2535 62
NM_001259854.2 NP_001246783.1 12 - 13 3620 62
NM_001259855.2 NP_001246784.1 13 - 14 3520 62
NM_001259856.2 NP_001246785.1 13 - 14 3776 62
NM_001259857.2 NP_001246786.1 13 - 14 3523 62
NM_001259858.2 NP_001246787.1 11 - 12 2591 62
NM_001274990.1 NP_001261919.1 11 - 12 2983 62
NM_001382059.1 NP_001369057.1 11 - 12 2983 62
NM_168646.3 NP_730099.2 12 - 13 3105 62
NM_168647.3 NP_730100.2 11 - 12 2347 62
NM_168648.3 NP_730101.2 11 - 12 2958 62
GenBank mRNA AF500094.1 - 10 - 11 2490 62
BT133053.1 - 12 - 13 3093 62

View Full Product Details