Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mbs
Entrez Gene ID
49070
Gene Name
Myosin binding subunit
Gene Aliases
0573/06, 0953/04, CG32156, CG5600, CG5891, DMBS, DMYPT, DMbs, Dmel\CG32156, Dmel_CG32156, MBS, Mypt, l(3)03802, l(3)72Dd, l(3)S005331b, l(3)S057306, l(3)S057306b, l(3)S095304, l(3)j3B4, l(3)j7A1, mbs
Chromosome Location
Chr.3L: 16051959 - 16082026 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005536
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104143.3 NP_001097613.1 6 - 7 1136 73
NM_001259854.2 NP_001246783.1 18 - 19 4349 73
NM_001259857.2 NP_001246786.1 19 - 20 4252 73
NM_001382055.1 NP_001369053.1 5 - 6 1601 73
NM_168646.3 NP_730099.2 18 - 19 3834 73
NM_168648.3 NP_730101.2 17 - 18 3687 73
GenBank mRNA AF500094.1 - 16 - 17 3219 73
BT133053.1 - 18 - 19 3822 73

View Full Product Details