Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rtc1
Entrez Gene ID
32338
Gene Name
CG11130-PA
Gene Aliases
CG11130, Dmel\CG11130, Dmel_CG11130
Chromosome Location
Chr.X: 13757179 - 13758598 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020909
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_132691.4 NP_572919.1 1 - 2 898 66
GenBank mRNA AY058762.1 - 1 - 2 909 66

View Full Product Details