Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rab3-GEF
Entrez Gene ID
32442
Gene Name
Rab3 GDP-GTP exchange factor
Gene Aliases
BEST:GH21304, CG5627, Dmel\CG5627, Dmel_CG5627, anon-EST:Posey246, rab3-GEF, rab3-gef
Chromosome Location
Chr.X: 15089682 - 15113762 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030613
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103513.3 NP_001096983.2 13 - 14 4635 79
NM_001258746.2 NP_001245675.1 14 - 15 4653 79
NM_001347810.1 NP_001334680.1 15 - 16 4695 79
NM_001347811.1 NP_001334681.1 15 - 16 4695 79

View Full Product Details