Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- shtd
- Entrez Gene ID
-
32473
- Gene Name
- shattered
- Gene Aliases
-
APC1, APC1/shtd, Apc1, CG9198, Dmel\CG9198, Dmel_CG9198, SHTD, Shtd
- Chromosome Location
-
Chr.X: 15455544 - 15462441 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004391
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132797.3 |
NP_573025.2 |
4 - 5 |
2655 |
89 |
View Full Product Details