Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSBP
Entrez Gene ID
32548
Gene Name
membrane steroid binding protein
Gene Aliases
CG9066, Dmel\CG9066, Dmel_CG9066, dm_MSBP
Chromosome Location
Chr.X: 15887567 - 15890109 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030703
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169294.2 NP_001162765.1 3 - 4 567 65
NM_001272658.1 NP_001259587.1 2 - 3 650 65
NM_132859.4 NP_573087.1 2 - 3 634 65
GenBank mRNA AB290904.1 - 2 - 3 603 65
AB290905.1 - 3 - 4 536 65
AY061163.1 - 2 - 3 617 65

View Full Product Details