Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSBP
Entrez Gene ID
32548
Gene Name
membrane steroid binding protein
Gene Aliases
CG9066, Dmel\CG9066, Dmel_CG9066, dm_MSBP
Chromosome Location
Chr.X: 15887567 - 15890109 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0030703
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169294.2 NP_001162765.1 4 - 5 709 69
NM_001272658.1 NP_001259587.1 3 - 4 792 69
NM_132859.4 NP_573087.1 3 - 4 776 69
GenBank mRNA AB290904.1 - 3 - 4 745 69
AB290905.1 - 4 - 5 678 69
AY061163.1 - 3 - 4 759 69

View Full Product Details