Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- dpr18
- Entrez Gene ID
-
32572
- Gene Name
- defective proboscis extension response 18
- Gene Aliases
-
CG14948, CT34788, Dmel\CG14948, Dmel_CG14948, Dpr-18, Dpr18
- Chromosome Location
-
Chr.X: 16063738 - 16070304 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0030723
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_132874.2 |
NP_573102.1 |
3 - 4 |
610 |
106 |
GenBank mRNA |
BT003521.1 |
- |
3 - 4 |
607 |
106 |
View Full Product Details