Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGRP-SD
- Entrez Gene ID
-
38858
- Gene Name
- Peptidoglycan recognition protein SD
- Gene Aliases
-
CG7496, Dm PGRP-SD, Dmel\CG7496, Dmel_CG7496, pgrp-sd
- Chromosome Location
-
Chr.3L: 7651180 - 7651900 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035806
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139888.3 |
NP_648145.1 |
1 - 1 |
540 |
67 |
View Full Product Details