Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- eIF4E5
- Entrez Gene ID
-
38878
- Gene Name
- eukaryotic translation initiation factor 4E5
- Gene Aliases
-
CG8277, CT24507, Dmel\CG8277, Dmel_CG8277, eIF4E, eIF4E-5
- Chromosome Location
-
Chr.3L: 7894880 - 7896009 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0035823
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139903.2 |
NP_648160.2 |
2 - 3 |
700 |
85 |
View Full Product Details