Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sarm
Entrez Gene ID
38895
Gene Name
Sterile alpha and Armadillo motif
Gene Aliases
CG13680, CG13681, CG34373, CG43119, CG7905, CG7915, Dmel\CG43119, Dmel_CG43119, Ect4, Ect4/SARM, SARM, anon-WO0118547.272, dSARM, dSARM1, dSarm, dsarm, ect4
Chromosome Location
Chr.3L: 8063874 - 8108897 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262579
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043129.3 NP_001036594.1 8 - 9 4852 83
NM_001274627.1 NP_001261556.1 10 - 11 5958 83
NM_001382096.1 NP_001369041.1 6 - 7 3744 83
GenBank mRNA BT014666.1 - 9 - 10 4732 83

View Full Product Details