Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sbp2
Entrez Gene ID
38934
Gene Name
SECIS-binding protein 2
Gene Aliases
CG7066, Dmel\CG7066, Dmel_CG7066, SBP2
Chromosome Location
Chr.3L: 8304157 - 8305325 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0087039
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139947.3 NP_648204.1 1 - 2 531 117
GenBank mRNA AY070491.1 - 1 - 2 528 117

View Full Product Details