Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhea
Entrez Gene ID
38978
Gene Name
CG6831-PB
Gene Aliases
CG6831, CT21159, Dmel\CG6831, Dmel_CG6831, Rhea, Talin, Tln, talin, tendrils
Chromosome Location
Chr.3L: 8545001 - 8574409 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260442
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259745.2 NP_001246674.1 7 - 8 1766 76
NM_001259746.2 NP_001246675.1 7 - 8 1766 76
NM_001259747.2 NP_001246676.1 7 - 8 1878 76
NM_001274657.1 NP_001261586.1 6 - 7 2119 76
NM_001274658.1 NP_001261587.1 7 - 8 1766 76
NM_001274659.1 NP_001261588.1 7 - 8 1888 76
NM_001274660.1 NP_001261589.1 7 - 8 1878 76
NM_139981.4 NP_648238.1 6 - 7 1591 76
GenBank mRNA AF299248.1 - 5 - 6 1538 76

View Full Product Details