Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Khc
Entrez Gene ID
36810
Gene Name
Kinesin heavy chain
Gene Aliases
CG7765, DK, DKH, Dm KHC, DmK, DmKHC, Dmel\CG7765, Dmel_CG7765, Dmkin, KHC, KIF 5A, KIF5, KIF5B, KIN, Kif5, Kin, Kin-1, Kinesin-1, khc, kin, kinesin, kinesin-1, l(2)W12, l(2)k13219, l(2)k13314, l(2R)W12, pgs
Chromosome Location
Chr.2R: 16266960 - 16271971 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001308
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057242.5 NP_476590.1 3 - 4 2978 76

View Full Product Details