Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dre4
- Entrez Gene ID
-
38248
- Gene Name
- CG1828-PA
- Gene Aliases
-
CG1828, DRE4, DRE4/dSPT16, Dmel\CG1828, Dmel_CG1828, Dre4, SPT16, Spt16, dSPT16, dSpt16, dspt16, l(3)62Bg, l(3)dre4, spt16
- Chromosome Location
-
Chr.3L: 1871574 - 1876312 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002183
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057262.4 |
NP_476610.2 |
8 - 9 |
3348 |
69 |
View Full Product Details