Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nub
Entrez Gene ID
34669
Gene Name
nubbin
Gene Aliases
CG15488, CG15489, CG34395, CG6246, Dmel\CG34395, Dmel_CG34395, NUB, Nb, Nub, Oct-1, Oct1, PDM-1, PDM1, POU33F1, Pdm, Pdm-1, Pdm1, dOct1, dPOU-19, dPOU-19/pdm-1, dPOU19, nb, pdm, pdm-1, pdm1, twn
Chromosome Location
Chr.2L: 12587625 - 12628135 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085424
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103683.2 NP_001097153.1 4 - 5 2056 60
NM_001298947.1 NP_001285876.1 2 - 3 388 60
NM_057311.5 NP_476659.1 2 - 3 826 60
GenBank mRNA AY118387.1 - 2 - 3 387 60
M65015.1 - 2 - 3 434 60
M81957.1 - 2 - 3 387 60

View Full Product Details