Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Sgs7
- Entrez Gene ID
-
47198
- Gene Name
- Salivary gland secretion 7
- Gene Aliases
-
68C GIII, CG18087, Dmel\CG18087, Dmel_CG18087, Sgs-7, sgs-7, sgs7
- Chromosome Location
-
Chr.3L: 11509861 - 11510247 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003377
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057370.2 |
NP_476718.1 |
1 - 2 |
70 |
63 |
View Full Product Details