Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cbp53E
Entrez Gene ID
36905
Gene Name
Calbindin 53E
Gene Aliases
CALB2, CALB2/calretinin, CG6702, Calb, Cbp33, DCBP, Dcbp, Dcp, Dmel\CG6702, Dmel_CG6702, cbn, cbp53E
Chromosome Location
Chr.2R: 16973927 - 16996057 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004580
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299582.1 NP_001286511.1 8 - 9 931 64
NM_057490.4 NP_476838.1 7 - 8 882 64
GenBank mRNA AY058340.1 - 7 - 8 849 64

View Full Product Details