Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ror
Entrez Gene ID
34367
Gene Name
CG4926-PA
Gene Aliases
CG4926, DRor, DmHD-2, Dmel\CG4926, Dmel_CG4926, Dror, HD-2, ROR, ROR1
Chromosome Location
Chr.2L: 10251843 - 10254686 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010407
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057614.3 NP_476962.1 1 - 2 227 93
GenBank mRNA BT125080.1 - 1 - 2 201 93
L20297.1 - 1 - 2 93

View Full Product Details