Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4e2
Entrez Gene ID
35822
Gene Name
Cytochrome P450-4e2
Gene Aliases
4e2, CG2060, CYP4E2, CyP4e2, Cyp4E2, Cyt-P450-4E2, Dmel\CG2060, Dmel_CG2060, P-450, P450, cyp4e2
Chromosome Location
Chr.2R: 8444560 - 8447164 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0014469
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299267.1 NP_001286196.1 5 - 6 1149 74
NM_057769.4 NP_477117.2 4 - 5 1052 74
GenBank mRNA AY058518.1 - 4 - 5 1028 74

View Full Product Details