Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
crol
Entrez Gene ID
34592
Gene Name
crooked legs
Gene Aliases
CG14938, Crol, Dmel\CG14938, Dmel_CG14938, ORE-15, anon-EST:Liang-1.74, anon-EST:Liang-2.48, clone 1.74, clone 2.48, croI, crol alpha, crol beta, crol gamma, l(2)04418, l(2)s2346
Chromosome Location
Chr.2L: 11794327 - 11809438 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020309
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259064.2 NP_001245993.1 4 - 5 2622 82
NM_001273464.1 NP_001260393.1 5 - 6 2086 82
NM_057895.4 NP_477243.1 5 - 6 2732 82
NM_057896.4 NP_477244.1 5 - 6 2114 82
NM_057897.4 NP_477245.1 5 - 6 2732 82
GenBank mRNA AF020347.1 - 5 - 6 2732 82
AF020348.1 - 5 - 6 2732 82
AF020349.1 - 5 - 6 2114 82

View Full Product Details