Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Idgf1
- Entrez Gene ID
-
34978
- Gene Name
- Imaginal disc growth factor 1
- Gene Aliases
-
BG:DS02780.5, CG4472, DmIDGF1, Dmel\CG4472, Dmel_CG4472, IDGF, IDGF-1, IDGF1, idgf1, ldgf1
- Chromosome Location
-
Chr.2L: 16445278 - 16446793 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020416
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057910.5 |
NP_477258.1 |
1 - 2 |
709 |
83 |
GenBank mRNA |
AY069157.1 |
- |
1 - 2 |
695 |
83 |
View Full Product Details