Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- rnh1
- Entrez Gene ID
-
35746
- Gene Name
- ribonuclease H1
- Gene Aliases
-
74/9, CG8729, Dmel\CG8729, Dmel_CG8729, RNase H1, RnH1, l(2)43F1-5, l(2)43Fa, l(2)k07409, l(2)k07624, rnhl
- Chromosome Location
-
Chr.2R: 7931054 - 7932380 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0023171
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_057967.5 |
NP_477315.1 |
1 - 2 |
373 |
67 |
GenBank mRNA |
AY089374.1 |
- |
1 - 2 |
379 |
67 |
View Full Product Details