Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dib
- Entrez Gene ID
-
45282
- Gene Name
- disembodied
- Gene Aliases
-
302a1, CG12028, CYP302A1, Cyp, Cyp27, Cyp302a1, Dib, Dmel\CG12028, Dmel_CG12028, l(3)64Ak, l(3)SH14, l(3)dib
- Chromosome Location
-
Chr.3L: 4062292 - 4064551 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000449
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080071.3 |
NP_524810.2 |
2 - 3 |
1200 |
68 |
GenBank mRNA |
BT029035.1 |
- |
2 - 3 |
1180 |
68 |
View Full Product Details