Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SuUR
Entrez Gene ID
45739
Gene Name
Suppressor of Under-Replication
Gene Aliases
CG7869, Dmel\CG7869, Dmel_CG7869, Dromel_CG7869_FBtr0076203_suur_mORF, SU(UR), SUUR, Su(UR), Su(UR)ES, su(UR)ES
Chromosome Location
Chr.3L: 11064991 - 11069025 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025355
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080096.3 NP_524835.1 3 - 4 1102 81
GenBank mRNA AJ277592.1 - 3 - 4 1079 81
BT010066.1 - 3 - 4 1078 81

Gene Symbol
CG45101
Entrez Gene ID
19834824
Gene Name
CG45101-PA
Gene Aliases
Dmel\CG45101, Dmel_CG45101, Dmel_SuUR_uORF
Chromosome Location
Chr.3L: 11064991 - 11069025 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266540
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300096.1 NP_001287025.1 3 - 4 1102 81

View Full Product Details