Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
S6KL
Entrez Gene ID
45970
Gene Name
S6 Kinase Like
Gene Aliases
3-10, BIN4, BcdK, Bin4, CG7001, Dmel\CG7001, Dmel_CG7001, PK17E, Pk17E, Pk?4, bin4, pk17e
Chromosome Location
Chr.X: 18788464 - 18806057 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283473
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258813.1 NP_001245742.1 8 - 9 1230 90
NM_001258814.2 NP_001245743.1 8 - 9 1702 90
NM_080122.4 NP_524861.2 8 - 9 1708 90
GenBank mRNA AY069491.1 - 8 - 9 1602 90

View Full Product Details