Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
east
Entrez Gene ID
46006
Gene Name
enhanced adult sensory threshold
Gene Aliases
CG4399, Dmel\CG4399, Dmel_CG4399, EAST, EG:133E12.4, East, anon-WO03040301.224, l(1)G0014, l(1)G0500, su(w[sp])
Chromosome Location
Chr.X: 2010860 - 2024647 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261954
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201601.2 NP_001188530.1 6 - 7 4553 72
NM_001201602.2 NP_001188531.1 6 - 7 4611 72
NM_001258555.1 NP_001245484.1 6 - 7 4749 72
NM_001258556.2 NP_001245485.1 6 - 7 4663 72
NM_001297869.1 NP_001284798.1 6 - 7 5205 72
NM_080125.5 NP_524864.2 6 - 7 4619 72

View Full Product Details