Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tutl
Entrez Gene ID
46015
Gene Name
turtle
Gene Aliases
CG15426, CG15427, CG15428, CT35489, CT35490, Dmel\CG15427, Dmel_CG15427, Tutl, jf4, l(2)01085, l(2)24Df, l(2)jf4, mdcds_7441
Chromosome Location
Chr.2L: 4283148 - 4323781 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0010473
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169397.2 NP_001162868.1 3 - 4 881 54
NM_001169398.1 NP_001162869.1 3 - 4 829 54
NM_001273090.1 NP_001260019.1 3 - 4 939 54
NM_001273091.1 NP_001260020.1 3 - 4 881 54
NM_001316378.1 NP_001303307.1 3 - 4 881 54
NM_001316379.1 NP_001303308.1 3 - 4 881 54
NM_080127.6 NP_524866.4 3 - 4 881 54
NM_164576.4 NP_722967.2 3 - 4 881 54
NM_164577.3 NP_722968.1 3 - 4 881 54
NM_164578.4 NP_722969.1 3 - 4 881 54
GenBank mRNA AJ312132.1 - 3 - 4 836 54

View Full Product Details