The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272577.2 | NP_001259506.1 | 4 - 5 | 2166 | 67 |
NM_080138.4 | NP_524877.1 | 4 - 5 | 2166 | 67 | |
GenBank mRNA | BT010259.1 | - | 4 - 5 | 2191 | 67 |